Order Entry
Poland
ContactUsLinkComponent
 
Cadmium(II) bromide, anhydrous 98%, pure
 
undefined
Cadmium(II) bromide, anhydrous 98%, pure

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to light yellow Crystalline powder
  • Assay:
    ≥97.5 %

 

 

Frequently Bought Together