N-Boc-1,3-propanediamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N-(3-Aminopropyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(3-Aminopropyl)carbamate, N-Boc-1,3-propanediamine, tert-Butyl (3-aminopropyl)carbamate, N-1-Boc-1,3-diaminopropane·HCl, N-(tert-Butoxycarbonyl)-1,3-propanediamine, N-Boc-1,3-diaminopropane
CAS Number:75178-96-0
:
Formula:C₈H₁₈N₂O₂
MDL Number:MFCD00210021
Boiling Pt:94…96 °C (.2Torr)
Melting Pt:22 °C
Density:0.998 g/cm³
Molecular Weight:174.244 g/mol
Storage Temperature:Ambient
UN:3259
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to white Liquid or low melting solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together