Order Entry
Poland
ContactUsLinkComponent
Calcium hydrogen orthophosphate dihydrate 98+%, extra pure
Calcium hydrogen orthophosphate dihydrate 98+%, extra pure
Certificates
undefined
Calcium hydrogen orthophosphate dihydrate 98+%, extra pure
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White to almost white Powder
  • Titration Complexometric:
    98.0 to 105.0 % (on dried substance)
  • Identification:
    Calcium: positive
  • Loss on ignition:
    24.5 % to 26.5 % (800°C)