2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-[N,N-Bis(Trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)triflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonyl)imide, Comins Triflating Reagent, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonimide)
CAS Number:145100-51-2
:
Formula:C₇H₃ClF₆N₂O₄S₂
MDL Number:MFCD00191833
Boiling Pt:77…90 °C (.2 mmHg)
Molecular Weight:392.67 g/mol
Melting Pt:45…47 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline solid
- Infrared spectrum:Conforms
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together