Disposable macro cuvettes
Supplier: Brand
Certificates
About this item
Plastic disposable UV-cuvettes for the UV/Visible range.
- Clear and clean optical path
- Manufactured under controlled room conditions
- UV transparent alternative to fragile glass or quartz cuvettes
Suitable for many applications for fluorescence spectroscopy.
Packaging: Automatically packed (without human contact) and grouped by mould cavity number to ensure lowest variation of extinction coefficient.
Frequently Bought Together