2,2,2-trifluoroethylhydrazine 70% 70% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (2,2,2-Trifluoroethyl)hydrazine, 2,2,2-Trifluoroethylhydrazine
CAS Number:5042-30-8
:
Formula:C₂H₅F₃N₂
MDL Number:MFCD00007622
Density:1.294 g/cm³
Molecular Weight:114.071 g/mol
Flash Pt:42 °C
Storage Temperature:Ambient
UN:1993
ADR:3 III
:GHS07
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Solution
- Refractive index:1.3600 to 1.3640 (20°C, 589 nm)
- Titration with HClO4:≥65 %
Specification Test Results
Frequently Bought Together