Order Entry
Poland
ContactUsLinkComponent
Amberlyst® 15, wet ion-exchange resin
Amberlyst® 15, wet ion-exchange resin
Certificates
undefined
Amberlyst® 15, wet ion-exchange resin
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Grey-brown Beads
  • Particle size:
    ≤1.0 % (< 0.355 mm)
  • Harmonic mean size:
    0.600 to 0.850 mm
  • Uniformity coefficient:
    ≤1.70
  • Moisture holding capacity:
    52.0 to 57.0 % (H)
  • Total volume capacity:
    ≥1.70 eq/l (H)
  • Total weight capacity:
    ≥4.70 eq/kg (H)