1-Phenylcyclobutanecarbonitrile 95%
ACRO154950250
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5313 to 1.5333 (20°C, 589 nm)
- GC:≥94 %
- NMR (1H-NMR):Consistent with structure
:
:157.216 g/mol
:3276
:GHS07
:Ambient
:14377-68-5
:C₁₁H₁₁N
:MFCD00019258
:1.03 g/cm³
:6.1 III