L-Tyrosine methyl ester 98%
: H-Tyr-OMe
:1080-06-4
:
:C₁₀H₁₃NO₃
:MFCD00002392
:195.22 g/mol
:133…135 °C
:GHS07
:Refrigerator
- Environmentally Preferable:
- Appearance:White to light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Appearance of solution:(5% soln. in methanol) Clear to slightly hazy
- Specific optical rotation:+23° to +29° (20°C, 589 nm) (c=2.4, CH3OH)
Frequently Bought Together