Order Entry
Poland
ContactUsLinkComponent
Anti-MX1 Mouse Monoclonal Antibody [clone: 474CT4.1.5]
Anti-MX1 Mouse Monoclonal Antibody [clone: 474CT4.1.5]
  ABGEAM2061B
 :  Abgent
Anti-MX1 Mouse Monoclonal Antibody [clone: 474CT4.1.5]
  ABGEAM2061B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    MX dynamin-like GTPase 1
  • Antigen symbol:
    MX1
  • Clonality:
    Monoclonal
  • Clone:
    474CT4.1.5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    4599
  • Antigen synonyms:
    MX|MxA|IFI-78K|IFI78
  • Amino acid number:
    617 to 646
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    75.52 kDa
  • Sequence:
    ERSDTSDKRKFLKERC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Shipping temperature:
    4 °C
  • Immunogen:
    This MX1 antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 617-646 amino acids from human MX1.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: MX1
Clonality: Monoclonal
Clone: 474CT4.1.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human