Specifications
- Antibody type:Primary
- Antigen name:Carboxypeptidase X (M14 Family), Member 1
- Antigen symbol:CPXM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:56265
- Antigen synonyms:CPX1|CPXM
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
CPXM (carboxypeptidase X, member 1) belongs to the peptidase M14 family. However, no carboxypeptidase activity has yet been detected. It may be involved in cell-cell interactions.Members of the M14 metallocarboxypeptidase protein family serve many diverse functions and are divided into three subfamilies based on structure, function and amino acid sequence similarity. Belonging to the N/E subfamily, CPXM (metallocarboxypeptidase CPX-1) is a 734 amino acid protein that contains a F5/8 type C domain and likely binds one zinc ion per subunit. Most members of the N/E subfamily contain several domains, including an active carboxypeptidase domain and signal peptide, and are thought to function mostly in protein-protein interactions and/or protein-membrane interactions, thereby targeting the protein to specific locations within the secretory pathway. CPXM is a unique member of this subfamily in that it does not appear to exhibit any enzymatic activity due to lack of several active-site residues that are present in the catalytic domain of other members of the N/E subfamily. Studies showing that CPXM expression is regulated during osteoclastogenesis suggest that CPXM may play a role in osteoclast differentiation. There are two isoforms of CPXM which are a result of alternative splicing events.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CPXM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: