Order Entry
Poland
ContactUsLinkComponent
 
Anti-SDH/Sorbitol Dehydrogenase Rabbit Polyclonal Antibody
  BOSSBS-3829R
 :  Bioss
Anti-SDH/Sorbitol Dehydrogenase Rabbit Polyclonal Antibody
  BOSSBS-3829R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Sorbitol Dehydrogenase
  • Antigen symbol:
    SDH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Gene ID:
    6390
  • Antigen synonyms:
    SORD1|LIR-7|CD85H|ILT1|LIR7|HEL-S-95n
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Sorbitol dehydrogenase (SDH), a member of the medium-chain dehydrogenase/reductase protein family and the second enzyme of the polyol pathway of glucose metabolism, converts sorbitol to fructose strictly using NAD(+) as coenzyme. SDH is expressed almost ubiquitously in all mammalian tissues. The enzyme has attracted considerable interest due to its implication in the development of diabetic complications as the polyol pathway is particularly active in hyperglycemic states. Although SORD is closely related to the class I long-chain alcohol dehydrogenases, it differs in substrate specificity, catalyzing polyols such as sorbitol and xylitol but having no activity towards primary alcohols.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SDH/Sorbitol Dehydrogenase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat