Order Entry
Poland
ContactUsLinkComponent
Anti-PNMA1 Antibody Pair
Anti-PNMA1 Antibody Pair
Catalog # ABNOH00009240-AP11
Supplier:  Abnova
Anti-PNMA1 Antibody Pair
Catalog # ABNOH00009240-AP11
Supplier:  Abnova

Specifications

  • Antigen name:
    Paraneoplastic Ma Antigen 1
  • Antigen symbol:
    PNMA1
  • Host:
    Multiple
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    9240
  • Antigen synonyms:
    37 kDa neuronal protein|Paraneoplastic antigen MA1|MA1
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Set comes with matched Antibody Pair to detect and quantify protein level of human PNMA1.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-PNMA1 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-PNMA1, IgG2a kappa (20 µg);

Type:
Antigen: PNMA1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human