Anti-GST tag Monoclonal Antibody
Catalog # ABNOMAB0042-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:GST tag
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Storage buffer:1x PBS, pH 7,4
- Sequence:MESPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDLEVLFQGPLEDPGYRGRTSFV
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:GST tag. MW of the GST tag is 26 KDa.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against GST recombinant protein.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: GST tag
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: