Order Entry
Poland
ContactUsLinkComponent
Anti-SIT1 Mouse Monoclonal Antibody [clone: SIT-01]
Anti-SIT1 Mouse Monoclonal Antibody [clone: SIT-01]
Catalog # ABNOMAB0933
Supplier:  Abnova
Anti-SIT1 Mouse Monoclonal Antibody [clone: SIT-01]
Catalog # ABNOMAB0933
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Signaling threshold regulating transmembrane adaptor 1
  • Antigen symbol:
    SIT1
  • Clonality:
    Monoclonal
  • Clone:
    SIT-01
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    27240
  • Antigen synonyms:
    SIT|SIT1_HUMAN|SHP intracting transmembrane adapter protein|Signaling threshold-regulating transmembrane adapter 1|gp30/40|SHP2-interacting transmembrane adapter protein|Sit1|Suppression-inducing transmembrane adapter 1.|Suppression inducing transmembrane adapter 1
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to intracellular region of human SIT1.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant SIT1.

Type: Primary
Antigen: SIT1
Clonality: Monoclonal
Clone: SIT-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human