Anti-FCGR3B Mouse Monoclonal Antibody [clone: MEM-154]
ABNOMAB0879
: Abnova
- Antibody type:Primary
- Antigen name:Fc fragment of IgG, low affinity 3B, receptor (CD16b)
- Antigen symbol:FCGR3B
- Clonality:Monoclonal
- Clone:MEM-154
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:2215
- Antigen synonyms:CD16b|CD16|FCRIII|FCGR3|FCG3|FCR-10|FCRIIIb
- Storage buffer:TBS, pH 8.0 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified FCGR3B from human granulocytes.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native FCGR3B.
Type: Primary
Antigen: FCGR3B
Clonality: Monoclonal
Clone: MEM-154
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human