Anti-TPX2 Rabbit Polyclonal Antibody
ABNOPAB11993
: Abnova
- Antibody type:Primary
- Antigen name:TPX2, Microtubule Nucleation Factor
- Antigen symbol:TPX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Mammals,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Antiserum
- Gene ID:22974
- Antigen synonyms:DIL-2|C20orf1|HCTP4|DIL2|C20orf2|p100|GD:C20orf1|FLS353|REPP86|HCA519
- Storage temperature:Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to C-terminus of human TPX2.
- Pk:100 µl
Rabbit polyclonal antibody raised against partial recombinant TPX2.
Recommended Dilutions: Immunofluorescence: 1:1000; Immunoprecipitation: 1:1000; Western Blot: 1:1000
Type: Primary
Antigen: TPX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mammals, Mouse, Rat