Order Entry
Poland
ContactUsLinkComponent
Anti-VAMP3 Mouse Monoclonal Antibody [clone: k2A2]
Anti-VAMP3 Mouse Monoclonal Antibody [clone: k2A2]
Catalog # ABNOMAB2061
Supplier:  Abnova
Anti-VAMP3 Mouse Monoclonal Antibody [clone: k2A2]
Catalog # ABNOMAB2061
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vesicle-Associated Membrane Protein 3
  • Antigen symbol:
    VAMP3
  • Clonality:
    Monoclonal
  • Clone:
    k2A2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    9341
  • Antigen synonyms:
    Synaptobrevin-3|VAMP-3|VAMP3|CEB|Cellubrevin|SYB3
  • Amino acid number:
    1 to 77
  • Storage buffer:
    PBS, pH 7,4 (0,1% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 1-77 of human VAMP3.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant VAMP3.

Type: Primary
Antigen: VAMP3
Clonality: Monoclonal
Clone: k2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse