Anti-NOL3 Rabbit Polyclonal Antibody
ABNOPAB13079
: Abnova
- Antibody type:Primary
- Antigen name:Nucleolar protein 3
- Antigen symbol:NOL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:8996
- Antigen synonyms:FCM|NOP|Apoptosis repressor with CARD|Myp|MYP|NOP30|Nucleolar protein 3|ARC
- Amino acid number:191 to 208
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 191-208 of human NOL3.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of NOL3.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: NOL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human