Order Entry
Poland
ContactUsLinkComponent
Anti-ARL4 Goat Polyclonal Antibody
Anti-ARL4 Goat Polyclonal Antibody
Catalog # ABNOPAB6217
Supplier:  Abnova
Anti-ARL4 Goat Polyclonal Antibody
Catalog # ABNOPAB6217
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADP-Ribosylation Factor-Like 4
  • Antigen symbol:
    ARL4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    10124
  • Antigen synonyms:
    ADP-ribosylation factor-like protein 4A|ADP ribosylation factor like 4A|ARL4A_HUMAN.|ARL4A|ARL 4
  • Storage buffer:
    Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
  • Sequence:
    GNGLSDQTSILSN-C
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human ARL4A.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Goat polyclonal antibody raised against synthetic peptide of ARL4A.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: ARL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human