Anti-ADD2 Rabbit Polyclonal Antibody
ABNOPAB18059
: Abnova
- Antibody type:Primary
- Antigen name:Adducin 2 Beta
- Antigen symbol:ADD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:119
- Antigen synonyms:ADDB
- Storage buffer:PBS, 150mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human ADD2.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of ADD2.
Recommended Dilutions: Western Blot: 1:500~1:1000; Immunofluorescence: 1:500~1:1000; ELISA: 1:5000
Type: Primary
Antigen: ADD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat