Order Entry
Poland
ContactUsLinkComponent
Anti-MMP3 Rabbit Polyclonal Antibody
Anti-MMP3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19143
Supplier:  Abnova
Anti-MMP3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19143
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 3
  • Antigen symbol:
    MMP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Marmoset,
    Rabbit,
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    4314
  • Antigen synonyms:
    STR1|transin-1|MMP-3|proteoglycanase|CHDS6|STMY|SL-1|matrix metalloproteinase 3|STMY1
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Purified human MMP3.
  • Purification:
    Protein A purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against MMP3.

Recommended Dilutions: Western Blot: 200-900 ng/ml; Immunoprecipitation: 10-30 ?g/ml

Type: Primary
Antigen: MMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Marmoset, Rabbit