Order Entry
Poland
ContactUsLinkComponent
Anti-IL27RA Rabbit Polyclonal Antibody
Anti-IL27RA Rabbit Polyclonal Antibody
  ABGEAP20662C
 :  Abgent
undefined
Anti-IL27RA Rabbit Polyclonal Antibody
  ABGEAP20662C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    interleukin 27 receptor, alpha
  • Antigen symbol:
    IL27RA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Gene ID:
    9466
  • Antigen synonyms:
    WSX1|zcytor1|IL27R|CRL1|TCCR|IL-27RA
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    69.474 kDa
  • Sequence:
    PHMEQVPEAQPLGD
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    1.000
  • Shipping temperature:
    4 °C
  • Immunogen:
    This IL27RA antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 569-602 amino acids from the C-terminal region of human IL27RA.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: IL27RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human