Order Entry
Poland
ContactUsLinkComponent
 
Anti-TGM3 Rabbit Polyclonal Antibody
  ABGEAP17540B
 :  Abgent
Anti-TGM3 Rabbit Polyclonal Antibody
  ABGEAP17540B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    transglutaminase 3
  • Antigen symbol:
    TGM3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Gene ID:
    7053
  • Antigen synonyms:
    TGE
  • Amino acid number:
    539 to 567
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    76.632 kDa
  • Sequence:
    KISYAQYEKYLKSDC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    1.400
  • Shipping temperature:
    4 °C
  • Immunogen:
    This TGM3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 539-567 amino acids from the C-terminal region of human TGM3.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: TGM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human