Anti-AIF Rabbit Polyclonal Antibody
Catalog # CAYM160773-500
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:Allograft Inflammatory Factor 1
- Antigen symbol:AIF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Amino acid number:151 to 180
- Sequence:RARDPGARVLIVSEDPELPYMRPPLSKELW
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human AIF amino acids 151-180
- Purification:purified by affinity chromatography
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: AIF
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat