Anti-ARO Rabbit Polyclonal Antibody
ORIGTA321053
: OriGene
- Antibody type:Primary
- Antigen symbol:ARO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Amino acid number:240 to 255
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:58 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to a region derived from 240-255 amino acids of Human Aromatase
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:50-200
Type: Primary
Antigen: ARO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat