Anti-Protein G Antibody (Biotin)
Catalog # BSBTBA1026-1
Supplier: Boster Bio
Specifications
- Antibody type:Tag
- Antigen name:Protein G
- Conjugation:Biotin
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Tag
Antigen: Protein G
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity: