Anti-PNP Sheep Polyclonal Antibody
USBIP4350-11
- Antibody type:Primary
- Antigen name:purine nucleoside phosphorylase
- Antigen symbol:PNP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:1 to 289
- Storage buffer:PBS,trehalose.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: PNP (purine nucleoside phosphorylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human