Order Entry
Poland
ContactUsLinkComponent
 
Anti-MAP Kinase 1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  USBIM2351-80E-HRP
undefined
Anti-MAP Kinase 1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  USBIM2351-80E-HRP

 

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase 1
  • Antigen symbol:
    MAPK1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:250-1:500

Type: Primary
Antigen: MAP Kinase 1
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human