Anti-Glutaminase, Kidney Isoform, Mitochondrial Rabbit polyclonal antibody
USBIG7119-92C
- Antibody type:Primary
- Antigen name:Glutaminase, Kidney Isoform, Mitochondrial
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Form:liquid
- Amino acid number:516 to 545
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50; Immunofluorescence: 1:10-1:50; Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: Glutaminase, Kidney Isoform, Mitochondrial
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse