Order Entry
Poland
ContactUsLinkComponent
 
Anti-Complement Component C5a R Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 9H73]
  USBIC0012-01F
Anti-Complement Component C5a R Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 9H73]
  USBIC0012-01F

 

  • Antibody type:
    Primary
  • Antigen name:
    Complement Component C5a R
  • Clonality:
    Monoclonal
  • Clone:
    9H73
  • Conjugation:
    FAM (Carboxyfluorescein)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    lyophilized powder
  • Storage buffer:
    25ugantibody, 1ml saline, 0,5% BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~25ug/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

 

 

Type: Primary
Antigen: Complement Component C5a R
Clonality: Monoclonal
Clone: 9H73
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human