Order Entry
Poland
ContactUsLinkComponent
Anti-Red Fluorescent Protein, DsRed, rDsRed Chicken Polyclonal Antibody
Anti-Red Fluorescent Protein, DsRed, rDsRed Chicken Polyclonal Antibody
Catalog # USBI168473
Supplier:  US Biological
Anti-Red Fluorescent Protein, DsRed, rDsRed Chicken Polyclonal Antibody
Catalog # USBI168473
Supplier:  US Biological

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Red Fluorescent Protein, DsRed, rDsRed
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Chicken
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    0,02M potassium phosphate, 0,15M sodium chloride, pH 7,2, 0,01% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by filtration, delipidation, salt fractionation and extensive dialysis
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 1:10000; Western Blot: 1:1000-1:3000

Type: Tag
Antigen: Red Fluorescent Protein, DsRed, rDsRed
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: