Anti-EIF5A Mouse Monoclonal Antibody [clone: 1E7]
USBI126256
- Antibody type:Primary
- Antigen name:Eukaryotic Translation Initiation Factor 5A
- Antigen symbol:EIF5A
- Clonality:Monoclonal
- Clone:1E7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:66 to 154
- Storage buffer:PBS, pH 7,2.
- Sequence:GKKYEDICPSTHNMDVPNIKRNDYQLICIQDGYLSLLTETGEVREDLKLPEGELGKEIEGKYNAGEDVQVSVMCAMSEEYAVAIKPCK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: EIF5A (eukaryotic translation initiation factor 5A)
Clonality: monoclonal
Clone: 10000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human