Order Entry
Poland
ContactUsLinkComponent
Anti-EVPLL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-EVPLL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI035260-HRP
Supplier:  US Biological
undefined
Anti-EVPLL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI035260-HRP
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    envoplakin-like
  • Antigen symbol:
    EVPLL
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    internal
  • Form:
    liquid
  • Amino acid number:
    117 to 146
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: EVPLL (envoplakin-like)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human