Order Entry
Poland
ContactUsLinkComponent
Anti-HA tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 12CA5]
Anti-HA tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 12CA5]
Catalog # USBI031009-HRP
Supplier:  US Biological
undefined
Anti-HA tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 12CA5]
Catalog # USBI031009-HRP
Supplier:  US Biological

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    HA tag
  • Antigen symbol:
    HA
  • Clonality:
    Monoclonal
  • Clone:
    12CA5
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

Specifications

About this item

  • Recommended dilution: ELISA: 1:5000; Western Blot: 1:1000

Type: Tag
Antigen: HA tag (HA tag)
Clonality: Monoclonal
Clone: 12CA5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: