Anti-MCM4 Rabbit Polyclonal Antibody
BSBTPA1652
- Antibody type:Primary
- Antigen name:Minichromosome Maintenance Complex Component 4
- Antigen symbol:MCM4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:S. cerevisiae|KIAA4003|DNA replication licensing factor MCM 4|Minichromosome maintenance deficient 4|MCM 4 minichromosome maintenance deficient 4|Minichromosome maintenance|AU045576|mcdc21|P1 CDC21|hCdc21|P1CDC21|MGC33310|Homolog of S. pombe cell devision cycle 21|P1-CDC21|Minichromosome maintenance deficient 4 homolog S. cerevisiae|19G AI325074|4|mitotin S. cerevisiae|mKIAA4003|Minichromosome maintenance deficient S. cerevisiae 4|CDC54|Mcmd4|Minichromosome maintenance complex component 4|MCM4_HUMAN|CDC21|homolog of|CDC 21|MCM4|MCM4 minichromosome maintenance deficient 4|DNA replication licensing factor MCM4|CDC 54.|CDC21 homolog|hCdc 21|Minichromosome Maintenance 4|MCM 4
- Amino acid number:815 to 829
- Sequence:TPALKYQQLFEDIRG
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human MCM4 (815-829aa TPALKYQQLFEDIRG), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: MCM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat