Anti-KSR2 Mouse Monoclonal Antibody [clone: 5E9]
Catalog # ABNOH00283455-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Kinase suppressor of ras 2
- Antigen symbol:KSR2
- Clonality:Monoclonal
- Clone:5E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:283455
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: KSR2
Clonality: Monoclonal
Clone: 5E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human