Anti-TAGAP Mouse Polyclonal Antibody
ABNOH00117289-B01P
: Abnova
- Antibody type:Primary
- Antigen name:T-cell activation RhoGTPase activating Protein
- Antigen symbol:TAGAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:117289
- Antigen synonyms:ARHGAP47, FKSG15, IDDM21, TAGAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: TAGAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human