Order Entry
Poland
ContactUsLinkComponent
Anti-UBE2Q2 Mouse Polyclonal Antibody
Anti-UBE2Q2 Mouse Polyclonal Antibody
  ABNOH00092912-B01
 :  Abnova
undefined
Anti-UBE2Q2 Mouse Polyclonal Antibody
  ABNOH00092912-B01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin-conjugating enzyme E2R 2
  • Antigen symbol:
    UBE2Q2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    92912
  • Antigen synonyms:
    CDC34B|UBC3B|E2-CDC34B
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: UBE2Q2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human