Anti-ABCC11 Mouse Polyclonal Antibody
ABNOH00085320-B01P
: Abnova
- Antibody type:Primary
- Antigen name:ATP-binding cassette, Subfamily C (CFTR/MRP), Member 11
- Antigen symbol:ABCC11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:85320
- Antigen synonyms:WW|member 11|MRP8|sub-family C (CFTR/MRP)|ATP binding cassette subfamily C member 11|ATP-binding cassette|EWWD
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: ABCC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human