Anti-WNK2 Mouse Monoclonal Antibody [clone: 2A2]
ABNOH00065268-M02
: Abnova
- Antibody type:Primary
- Antigen name:WNK lysine deficient Protein kinase 2
- Antigen symbol:WNK2
- Clonality:Monoclonal
- Clone:2A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:65268
- Antigen synonyms:P/OKcl.13|SDCCAG43|NY-CO-43|PRKWNK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: WNK2
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human