Anti-NEUROD6 Mouse Monoclonal Antibody [clone: 3B3]
ABNOH00063974-M06
: Abnova
- Antibody type:Primary
- Antigen name:Neuronal differentiation 6
- Antigen symbol:NEUROD6
- Clonality:Monoclonal
- Clone:3B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:63974
- Antigen synonyms:Math-2|NEX1M|bHLHa2|Nex1|MATH2|Atoh2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NEUROD6
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human