Anti-NLGN4X Mouse Polyclonal Antibody
ABNOH00057502-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Neuroligin 4, X-linked
- Antigen symbol:NLGN4X
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:57502
- Antigen synonyms:ASPGX2, AUTSX2, HLNX, HNL4X, NLGN4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: NLGN4X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human