Order Entry
Poland
ContactUsLinkComponent
Anti-TCIRG1 Mouse Polyclonal Antibody
Anti-TCIRG1 Mouse Polyclonal Antibody
Catalog # ABNOH00010312-B01P
Supplier:  Abnova
undefined
Anti-TCIRG1 Mouse Polyclonal Antibody
Catalog # ABNOH00010312-B01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    T-cell, immune Regulator 1, ATPase, H+ transporting, lysosomal V0 Subunit A3
  • Antigen symbol:
    TCIRG1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    10312
  • Antigen synonyms:
    V-type proton ATPase 116 kDa subunit a isoform 3|TIRC7|OPTB1|ATP6N1C|OC116|Osteoclastic proton pump 116 kDa subunit|Stv1|OC-116kDa|a3|V-ATPase 116 kDa isoform a3|Atp6i|Vph1|ATP6V0A3|TCIRG1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: TCIRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human