Anti-CCRL2 Mouse Polyclonal Antibody
ABNOH00009034-B02P
: Abnova
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) receptor-like 2
- Antigen symbol:CCRL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9034
- Antigen synonyms:CKRX|CRAM-B|CRAM-A|HCR|CRAM|ACKR5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: CCRL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human