Anti-EIF3H Mouse Monoclonal Antibody [clone: 3B12]
Catalog # ABNOH00008667-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 3, subunit H
- Antigen symbol:EIF3H
- Clonality:Monoclonal
- Clone:3B12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8667
- Antigen synonyms:eIF3-gamma|eIF3-p40|EIF3S3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: EIF3H
Clonality: Monoclonal
Clone: 3B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human