Anti-VBP1 Rabbit Polyclonal Antibody
ABNOH00007411-D01
: Abnova
- Antibody type:Primary
- Antigen name:von Hippel-Lindau binding protein 1
- Antigen symbol:VBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7411
- Antigen synonyms:VBP-1|PFDN3|HIBBJ46|PFD3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: VBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human