Order Entry
Poland
ContactUsLinkComponent
 
Anti-TPMT Mouse Monoclonal Antibody [clone: 1B5]
  ABNOH00007172-M01
 :  Abnova
undefined
Anti-TPMT Mouse Monoclonal Antibody [clone: 1B5]
  ABNOH00007172-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Thiopurine S-methyltransferase
  • Antigen symbol:
    TPMT
  • Clonality:
    Monoclonal
  • Clone:
    1B5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    7172
  • Antigen synonyms:
    Thiopurine methyltransferase|S adenosyl L methionine thiopurine S methyltransferase|TPMT|HGNC:12014|Thiopurine S-methyltransferase|Thiopurine S methyltransferase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: TPMT
Clonality: Monoclonal
Clone: 1B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human