Anti-SKP1 Mouse Monoclonal Antibody [clone: 1H8]
ABNOH00006500-M01
: Abnova
- Antibody type:Primary
- Antigen name:S-phase Kinase-associated Protein 1
- Clonality:Monoclonal
- Clone:1H8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6500
- Antigen synonyms:transcription elongation factor B|p19A|SIII|EMC19|OCP-2|RNA polymerase II elongation factor-like protein OCP2|cyclin A/CDK2-associated protein p19|OCP-II|TCEB1L|organ of Corti protein II|organ of Corti protein 2|SKP1A|p19skp1|cyclin-A/CDK2-associated protein p19|OCP2|cyclin A/CDK2-associated p19
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SKP1
Clonality: Monoclonal
Clone: 1H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human