Anti-S100 A2 Mouse Monoclonal Antibody [clone: 2D10-A3]
Catalog # ABNOH00006273-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:S100 Calcium binding Protein A2
- Antigen symbol:S100 A2
- Clonality:Monoclonal
- Clone:2D10-A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6273
- Antigen synonyms:CAN19|S100L
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: S100 A2
Clonality: Monoclonal
Clone: 2D10-A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human